Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cope | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021538.1 | NP_067513.1 | 5 - 6 | 519 | 75 | |
GenBank mRNA | AB039837.1 | - | 5 - 6 | 519 | 75 | |
AK003458.1 | - | 5 - 6 | 536 | 75 | ||
AK140753.1 | - | 3 - 4 | 2240 | 75 | ||
BC009170.1 | - | 5 - 6 | 527 | 2812643 | 75 | |
BC057328.1 | - | 3 - 4 | 2205 | 75 | ||
BC083336.1 | - | 5 - 6 | 501 | 5709960 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |