Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hp1bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285480.1 | NP_001272409.1 | 2 - 3 | 226 | 79 | |
NM_001285481.1 | NP_001272410.1 | 3 - 4 | 424 | 79 | ||
GenBank mRNA | AK075725.1 | - | 2 - 3 | 204 | 79 | |
AK166161.1 | - | 3 - 4 | 410 | 79 | ||
BU612149.1 | - | 2 - 3 | 266 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |