Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kazn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144531.3 | NP_653114.3 | 12 - 13 | 2213 | 71 | |
GenBank mRNA | AK032460.1 | - | 10 - 11 | 1452 | 71 | |
AK173090.1 | - | 12 - 13 | 2213 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |