Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nktr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010918.2 | NP_035048.3 | 13 - 14 | 4162 | 72 | |
GenBank mRNA | AK144214.1 | - | 13 - 14 | 4073 | 72 | |
L04289.2 | - | 13 - 14 | 4167 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |