Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb6d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076790.2 | NP_001070258.1 | 3 - 4 | 546 | 121 | |
NM_001300892.1 | NP_001287821.1 | 3 - 4 | 391 | 121 | ||
GenBank mRNA | AK136270.1 | - | 3 - 4 | 548 | 121 | |
BC147559.1 | - | 3 - 4 | 391 | 9087441 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |