Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mau2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167939.1 | NP_001161411.1 | 3 - 4 | 431 | 64 | |
NM_028993.4 | NP_083269.4 | 3 - 4 | 431 | 64 | ||
GenBank mRNA | AK018662.1 | - | 3 - 4 | 399 | 64 | |
AK034837.1 | - | 3 - 4 | 393 | 64 | ||
AK039196.1 | - | 3 - 4 | 398 | 64 | ||
AK049277.1 | - | 3 - 4 | 392 | 64 | ||
AK081967.1 | - | 3 - 4 | 432 | 64 | ||
AK162318.1 | - | 3 - 4 | 398 | 64 | ||
AK164005.1 | - | 3 - 4 | 398 | 64 | ||
BC125568.1 | - | 3 - 4 | 431 | 40130083 | 64 | |
BC132139.1 | - | 3 - 4 | 431 | 40130416 | 64 | |
BC144990.1 | - | 3 - 4 | 431 | 9053538 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |