Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122829.1 | NP_001116301.1 | 2 - 3 | 637 | 84 | |
NM_030680.2 | NP_109605.2 | 2 - 3 | 637 | 84 | ||
GenBank mRNA | AF182947.1 | - | 3 - 4 | 626 | 84 | |
AF322655.1 | - | 2 - 3 | 357 | 84 | ||
AK148196.1 | - | 3 - 4 | 614 | 84 | ||
AK205256.1 | - | 1 - 2 | 148 | 84 | ||
BC052149.1 | - | 2 - 3 | 575 | 6332288 | 84 | |
BC056442.1 | - | 2 - 3 | 637 | 5702493 | 84 | |
BQ771453.1 | - | 2 - 3 | 637 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |