Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011078.3 | NP_035208.2 | 19 - 20 | 2826 | 71 | |
GenBank mRNA | AB221590.1 | - | 8 - 9 | 1212 | 71 | |
AF043726.1 | - | 19 - 20 | 2826 | 71 | ||
AK134358.1 | - | 5 - 6 | 654 | 71 | ||
AK172995.1 | - | 17 - 18 | 2533 | 71 | ||
BC051633.1 | - | 19 - 20 | 2824 | 6518591 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |