Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009654.3 | NP_033784.2 | 11 - 12 | 1475 | 68 | |
GenBank mRNA | AJ011413.1 | - | 11 - 12 | 1462 | 68 | |
AK010025.1 | - | 11 - 12 | 1464 | 68 | ||
AK050248.1 | - | 11 - 12 | 1464 | 68 | ||
AK050644.1 | - | 10 - 11 | 1333 | 68 | ||
AK131891.1 | - | 11 - 12 | 1467 | 68 | ||
AK160487.1 | - | 11 - 12 | 1467 | 68 | ||
BC024643.1 | - | 11 - 12 | 1449 | 4242076 | 68 | |
BC049971.1 | - | 11 - 12 | 1449 | 4161986 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |