Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK081904.1 | - | 1 - 2 | 214 | 63 | |
AK165116.1 | - | 1 - 2 | 214 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |