Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200017489.mm.1 |
NM_001142410.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017489.mm.1 |
NM_001142410.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017489.mm.1 |
TC0200002215.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200017490.mm.1 |
NM_001142410.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017490.mm.1 |
NM_001142410.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017490.mm.1 |
TC0200002215.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200017487.mm.1 |
NM_001142410.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017487.mm.1 |
NM_001142410.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017487.mm.1 |
TC0200002215.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200017488.mm.1 |
NM_001142410.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017488.mm.1 |
NM_001142410.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017488.mm.1 |
TC0200002215.mm.1 |
MTA TC |