Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2a22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101467.1 | NP_001094937.1 | 3 - 4 | 520 | 153 | |
GenBank mRNA | AK149509.1 | - | 3 - 4 | 519 | 153 | |
BC151116.1 | - | 3 - 4 | 514 | 9088029 | 153 | |
BC158034.1 | - | 3 - 4 | 514 | 9007337 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |