Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn2r60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105057.1 | NP_001098527.1 | 3 - 4 | 1310 | 120 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |