Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500002138.mm.1 |
TC0500000277.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500002138.mm.1 |
NM_001037940.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002138.mm.1 |
NM_001037940.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002138.mm.1 |
NM_001127367.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002138.mm.1 |
NM_001127367.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002138.mm.1 |
NM_011847.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002138.mm.1 |
NM_011847.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002148.mm.1 |
TC0500000277.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500002148.mm.1 |
NM_001037940.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002148.mm.1 |
NM_001037940.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002137.mm.1 |
TC0500000277.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500002137.mm.1 |
NM_001037940.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002137.mm.1 |
NM_001037940.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002137.mm.1 |
NM_001127367.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002137.mm.1 |
NM_001127367.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002137.mm.1 |
NM_011847.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002137.mm.1 |
NM_011847.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002142.mm.1 |
TC0500000277.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500002142.mm.1 |
NM_001037940.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002142.mm.1 |
NM_001037940.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002142.mm.1 |
NM_001127367.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002142.mm.1 |
NM_001127367.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002142.mm.1 |
NM_011847.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002142.mm.1 |
NM_011847.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002150.mm.1 |
TC0500000277.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500002150.mm.1 |
NM_001037940.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002150.mm.1 |
NM_001037940.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002140.mm.1 |
TC0500000277.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500002140.mm.1 |
NM_001037940.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002140.mm.1 |
NM_001037940.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002140.mm.1 |
NM_001127367.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002140.mm.1 |
NM_001127367.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002140.mm.1 |
NM_011847.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002140.mm.1 |
NM_011847.4 |
Refseq |