Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P111070 |
BC157977.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P111070 |
BC158124.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P111070 |
NM_001163288.2 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
633102 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
588634 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
406770 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
415066 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
83186 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
867672 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
150849 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
876438 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
403426 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
94814 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
561883 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
548445 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
552230 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
505222 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
941130 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
223588 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
514557 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
631279 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
48374 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
823572 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
430052 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
131404 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
148853 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
430112 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
400139 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
305195 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
233039 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
88376 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
648980 |
AK019538.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
648980 |
XM_001473955.1 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
648980 |
XM_001474001.1 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
648980 |
XM_001476324.1 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
648980 |
XM_136124.4 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
648980 |
XM_486800.5 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
648980 |
XM_486801.3 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
648980 |
XM_486805.3 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
648980 |
XM_621118.3 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
648980 |
XM_621121.4 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
648980 |
XM_910554.2 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
648980 |
XM_910573.2 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
648980 |
XM_910605.2 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
648980 |
XM_910844.2 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
648980 |
XM_989376.1 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
785725 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
169693 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
439675 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
939565 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
882258 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
489263 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
329192 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
689053 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
513360 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
725259 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
50795 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
273933 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
956322 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
484909 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
273839 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
670644 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
534904 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
759592 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
567355 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
401692 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
627527 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
289064 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
949562 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
133018 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
775242 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
884317 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
41514 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
547398 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
751735 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
625377 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
164685 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
34872 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
532929 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
805990 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
466729 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
701438 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
452450 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
879260 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
564809 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
143078 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
616470 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
642815 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
913880 |
AK002774.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
913880 |
AK011319.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
913880 |
AK135762.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
913880 |
AK135797.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
913880 |
AK162164.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
913880 |
AK162256.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
913880 |
BC016263.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
913880 |
BC085478.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
913880 |
NM_028106.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
734078 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
19478 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
856366 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
906999 |
AB041562.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
906999 |
AF053550.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
906999 |
AK083055.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
906999 |
AK087933.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
906999 |
AK168000.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
906999 |
AK168021.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
906999 |
AK168045.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
906999 |
AK169048.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
906999 |
BC006641.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
906999 |
NM_016804.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
91063 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
80900 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
269 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
40858 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
22599 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
108109 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
141372 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
128349 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
160073 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
105122 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
128386 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
12376 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
25847 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P111070 |
BC157977.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P111070 |
BC158124.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P111070 |
NM_001163288.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2149630 |
AK084362.1 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05253443 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P228073 |
AK041075.1 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P05253443 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955841 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22956059 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03969569 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955733 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03896374 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03221247 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03312684 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03347376 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22956019 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03621458 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03371208 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03244148 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955962 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03534312 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03937576 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03827346 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955845 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955756 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955982 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955919 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955783 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955996 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22956049 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03368037 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03680648 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955959 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955798 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955763 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955977 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955920 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03832145 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03827344 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955992 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22956063 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955741 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955717 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955917 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03937577 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03724431 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955730 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955839 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04029338 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22956053 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955921 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955792 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03756475 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22956002 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955734 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955924 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955969 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955801 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03773350 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955923 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955991 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955838 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04031273 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955980 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955771 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955789 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955970 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955978 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03765961 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955740 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955755 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03312683 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955835 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955752 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955914 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03271750 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955779 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03756474 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955799 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955803 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955918 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03724426 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22956020 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03984951 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955723 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22956057 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955725 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03724424 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22956036 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955811 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22956030 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03347375 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03885803 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955797 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955830 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22956043 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955848 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03904793 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03485838 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03485846 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03740490 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955953 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22956009 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03312682 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955820 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03290350 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03954904 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955853 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22956015 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03724423 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22956058 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22956010 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955764 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03621470 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955832 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955834 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955963 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03969572 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955759 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955780 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03504101 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955762 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955777 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955847 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22956001 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22956024 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03907769 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955710 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03671883 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955790 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955987 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955736 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955833 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22956047 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22956039 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03223215 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22956007 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22956028 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04010003 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955727 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955806 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03991312 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04100481 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04100482 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955990 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955956 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22956031 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955804 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03460751 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955809 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22956014 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955911 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04007160 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955719 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955973 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03546880 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22956027 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22956011 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955742 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955767 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955768 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955735 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03765963 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955998 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03621466 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955720 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955968 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03312681 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22956062 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22956056 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955976 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03312691 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03680643 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03685942 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22956004 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955807 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03621459 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03637875 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22956034 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22955837 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03984950 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03312676 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22956033 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03621465 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03621457 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22956051 |
- |
- |