Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900027632.mm.1 |
NM_028384.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027632.mm.1 |
NM_028384.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027632.mm.1 |
TC0900003273.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900027621.mm.1 |
NM_028384.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027621.mm.1 |
NM_028384.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027621.mm.1 |
TC0900003273.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900027624.mm.1 |
NM_028384.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027624.mm.1 |
NM_028384.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027624.mm.1 |
TC0900003273.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900027625.mm.1 |
NM_028384.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027625.mm.1 |
NM_028384.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027625.mm.1 |
TC0900003273.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900027628.mm.1 |
NM_028384.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027628.mm.1 |
NM_028384.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027628.mm.1 |
TC0900003273.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900027626.mm.1 |
NM_028384.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027626.mm.1 |
NM_028384.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027626.mm.1 |
TC0900003273.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900027627.mm.1 |
NM_028384.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027627.mm.1 |
NM_028384.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027627.mm.1 |
TC0900003273.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900027631.mm.1 |
NM_028384.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027631.mm.1 |
NM_028384.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027631.mm.1 |
TC0900003273.mm.1 |
MTA TC |