Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK046263.1 | - | 3 - 4 | 371 | 101 | |
BC138962.1 | - | 3 - 4 | 353 | 101 | ||
BC138963.1 | - | 3 - 4 | 353 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |