Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190451.1 | NP_001177380.1 | 2 | 440 | 153 | |
NM_007833.5 | NP_031859.1 | 2 | 348 | 153 | ||
GenBank mRNA | AK031456.1 | - | 1 | 2774 | 153 | |
AK135287.1 | - | 2 | 332 | 153 | ||
AK145905.1 | - | 2 | 330 | 153 | ||
AK147020.1 | - | 2 | 332 | 153 | ||
AK161788.1 | - | 2 | 330 | 153 | ||
AK165408.1 | - | 2 | 330 | 153 | ||
BC060126.1 | - | 1 | 161 | 153 | ||
BC132521.1 | - | 2 | 199 | 40130798 | 153 | |
BC138564.1 | - | 2 | 199 | 8861584 | 153 | |
BG088078.2 | - | 2 | 440 | 153 | ||
X53929.1 | - | 2 | 357 | 153 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon |