Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mprip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012027.2 | NP_036157.2 | 12 - 13 | 1696 | 84 | |
NM_201245.3 | NP_957697.1 | 12 - 13 | 1696 | 84 | ||
GenBank mRNA | AK035435.1 | - | 6 - 7 | 84 | ||
AK044512.1 | - | 11 - 12 | 1338 | 84 | ||
AK154807.1 | - | 13 - 14 | 1703 | 84 | ||
AK155630.1 | - | 12 - 13 | 1477 | 84 | ||
AK158296.1 | - | 12 - 13 | 1685 | 84 | ||
AK162530.1 | - | 10 - 11 | 1042 | 84 | ||
AK170849.1 | - | 12 - 13 | 1680 | 84 | ||
AK171089.1 | - | 12 - 13 | 1685 | 84 | ||
BC039932.1 | - | 11 - 12 | 1220 | 84 | ||
BC049803.1 | - | 12 - 13 | 1222 | 84 | ||
BC063067.1 | - | 12 - 13 | 1659 | 5721396 | 84 | |
U73200.1 | - | 12 - 13 | 1556 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |