Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sike1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK139055.1 | - | 1743 | 82 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Downstream |
cold shock domain containing E1, RNA binding |
Csde1 |
AA960392, BC016898, D3Jfr1, mKIAA0885, unr |
12.3kb | Sense | Chr. 3 - 103020431 - 103058189 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |