Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taco1os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK019365.1 | - | 1 - 2 | 43 | 60 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | DDB1 and CUL4 associated factor 7 | Dcaf7 |
1700012F10Rik, 2610037L01Rik, C86529, HAN11, Wdr68 |
-5.8kb | Anti-sense | Chr. 11 - 106036872 - 106059324 on NCBI Build GRCm38 |
Downstream | translational activator of mitochondrially encoded cytochrome c oxidase I | Taco1 |
2310066I18Rik, Ccdc44 |
107.0 | Anti-sense | Chr. 11 - 106066039 - 106073612 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Probe spans exons |