Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 9430019J16 gene | 9430019J16Rik | -14.8kb | Sense | Chr. 2 - 75436057 - 75444179 on NCBI Build GRCm38 | |
Downstream | heterogeneous nuclear ribonucleoprotein A3 | Hnrnpa3 |
2410013L13Rik, 2610209F03Rik, 2610510D13Rik, Hnrpa3 |
194.4kb | Anti-sense | Chr. 2 - 75659230 - 75669407 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |