Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ighd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | V00790.1 | - | 1 | 154 | 68 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Immunoglobulin heavy constant gamma 3 | Ighg3 | IgG3 | -48.2kb | Sense | Chr. 12 - 113357442 - 113361232 on NCBI Build GRCm38 |
Downstream | immunoglobulin heavy constant mu | Ighm |
AI326478, Igh-6, Igh-M, Igh6, Igm, muH |
9.2kb | Sense | Chr. 12 - 113418826 - 113422730 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |