Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF014450.1 | - | 1 | 247 | 94 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | tubulin-specific chaperone E | Tbce |
2610206D02Rik, C530005D02Rik, pmn |
-10.2kb | Sense | Chr. 13 - 13997949 - 14039638 on NCBI Build GRCm38 |
Downstream | AT rich interactive domain 4B (RBP1-like) | Arid4b |
BCAA, BRCAA1, RBBP1L1, Rbp1l1, SAP180 |
13.2kb | Anti-sense | Chr. 13 - 14063768 - 14199603 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon |