Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clrn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163317.1 | NP_001156789.1 | 1 - 2 | 307 | 72 | |
GenBank mRNA | BC147370.1 | - | 1 - 2 | 306 | 9056434 | 72 |
BC147371.1 | - | 1 - 2 | 306 | 9056432 | 72 | |
BC147726.1 | - | 1 - 2 | 307 | 9056675 | 72 | |
BC147731.1 | - | 1 - 2 | 307 | 9056680 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |