Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166031.2 | NP_001159503.2 | 3 - 4 | 264 | 71 | |
NM_080452.3 | NP_536700.3 | 3 - 4 | 321 | 71 | ||
GenBank mRNA | AB055389.1 | - | 3 - 4 | 312 | 71 | |
AK051184.1 | - | 3 - 4 | 321 | 71 | ||
AK077219.1 | - | 4 - 5 | 490 | 71 | ||
AK154131.1 | - | 3 - 4 | 315 | 71 | ||
AK160129.1 | - | 3 - 4 | 313 | 71 | ||
AK165956.1 | - | 3 - 4 | 312 | 71 | ||
AK172120.1 | - | 2 - 3 | 784 | 71 | ||
BC033460.1 | - | 2 - 3 | 770 | 71 | ||
BC060503.1 | - | 3 - 4 | 297 | 6306321 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |