Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200000315.mm.1 |
NM_001113558.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000315.mm.1 |
NM_001113558.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000315.mm.1 |
NM_001165932.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000315.mm.1 |
NM_001165932.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000315.mm.1 |
NM_026754.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000315.mm.1 |
NM_026754.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000315.mm.1 |
TC0200000040.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200000316.mm.1 |
NM_001113558.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000316.mm.1 |
NM_001113558.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000316.mm.1 |
NM_001165932.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000316.mm.1 |
NM_001165932.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000316.mm.1 |
NM_026754.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000316.mm.1 |
NM_026754.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000316.mm.1 |
TC0200000040.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200000318.mm.1 |
NM_001113558.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000318.mm.1 |
NM_001113558.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000318.mm.1 |
NM_001165932.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000318.mm.1 |
NM_001165932.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000318.mm.1 |
NM_026754.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000318.mm.1 |
NM_026754.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000318.mm.1 |
TC0200000040.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200000320.mm.1 |
NM_001113558.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000320.mm.1 |
NM_001113558.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000320.mm.1 |
NM_001165932.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000320.mm.1 |
NM_001165932.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000320.mm.1 |
NM_026754.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000320.mm.1 |
NM_026754.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000320.mm.1 |
TC0200000040.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200000317.mm.1 |
NM_001113558.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000317.mm.1 |
NM_001113558.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000317.mm.1 |
NM_001165932.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000317.mm.1 |
NM_001165932.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000317.mm.1 |
NM_026754.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000317.mm.1 |
NM_026754.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000317.mm.1 |
TC0200000040.mm.1 |
MTA TC |