Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ruvbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019685.2 | NP_062659.1 | 4 - 5 | 619 | 87 | |
GenBank mRNA | AF100694.1 | - | 4 - 5 | 658 | 87 | |
AK012170.1 | - | 4 - 5 | 567 | 87 | ||
AK132783.1 | - | 4 - 5 | 601 | 87 | ||
AK146258.1 | - | 4 - 5 | 611 | 87 | ||
AK146374.1 | - | 4 - 5 | 612 | 87 | ||
AK156078.1 | - | 4 - 5 | 589 | 87 | ||
BC004718.1 | - | 4 - 5 | 585 | 3582426 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |