Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rybp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019743.3 | NP_062717.2 | 4 - 5 | 510 | 78 | |
GenBank mRNA | AK033024.1 | - | 4 - 5 | 549 | 78 | |
BC053016.1 | - | 4 - 5 | 510 | 6406584 | 78 | |
BC080287.1 | - | 4 - 5 | 593 | 6410301 | 78 | |
BC138324.1 | - | 4 - 5 | 492 | 8861344 | 78 | |
BC138325.1 | - | 4 - 5 | 492 | 8861345 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |