Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pomp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025624.2 | NP_079900.1 | 1 - 2 | 67 | 85 | |
GenBank mRNA | AK003439.1 | - | 1 - 2 | 60 | 85 | |
AK011115.1 | - | 1 - 2 | 68 | 85 | ||
AK013934.1 | - | 1 - 2 | 61 | 85 | ||
AK167958.1 | - | 1 - 2 | 68 | 85 | ||
BC027016.1 | - | 1 - 2 | 58 | 5375452 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |