Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027462.4 | NP_081738.2 | 2 - 3 | 353 | 82 | |
GenBank mRNA | AK017602.1 | - | 2 - 3 | 364 | 82 | |
AK033774.1 | - | 351 | 82 | |||
AK036902.1 | - | 2 - 3 | 373 | 82 | ||
AK042715.1 | - | 2 - 3 | 354 | 82 | ||
AK042964.1 | - | 2 - 3 | 354 | 82 | ||
BC089583.1 | - | 2 - 3 | 375 | 6762967 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |