Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrnp35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029532.2 | NP_083808.1 | 1 - 2 | 87 | 61 | |
GenBank mRNA | AK039064.1 | - | 1 - 2 | 109 | 61 | |
BC043031.1 | - | 1 - 2 | 87 | 5693273 | 61 | |
BC058361.1 | - | 1 - 2 | 87 | 5693273 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |