Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145827.3 | NP_665826.1 | 1 - 2 | 178 | 104 | |
GenBank mRNA | AF486632.1 | - | 2 - 3 | 74 | 104 | |
AY495376.1 | - | 1 - 2 | 178 | 104 | ||
AY495377.1 | - | 2 - 3 | 74 | 104 | ||
BC116174.1 | - | 1 - 2 | 138 | 40048724 | 104 | |
BC116175.1 | - | 1 - 2 | 138 | 40048725 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |