Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146477.2 | NP_666688.2 | 1 | 551 | 101 | |
GenBank mRNA | BC119191.1 | - | 1 | 551 | 8733940 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |