Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172974.2 | NP_766562.1 | 1 - 2 | 130 | 96 | |
GenBank mRNA | AF071317.1 | - | 1 - 2 | 108 | 96 | |
AK053615.1 | - | 1 - 2 | 89 | 96 | ||
AK081309.1 | - | 1 - 2 | 130 | 96 | ||
AK168740.1 | - | 1 - 2 | 89 | 96 | ||
AK170140.1 | - | 1 - 2 | 89 | 96 | ||
BC012659.1 | - | 1 - 2 | 94 | 4017012 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |