Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk1b16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008454.2 | NP_032480.1 | 1 | 354 | 80 | |
GenBank mRNA | J03877.1 | - | 1 | 354 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |