Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mir30e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_029602.1 | 1 | 39 | 90 |
Gene Symbol: Mir30f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_105851.1 | 1 | 90 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | nuclear transcription factor-Y gamma | Nfyc | Sense | Chr. 4 - 120757435 - 120831579 on NCBI Build GRCm38 | ||
Downstream | regulating synaptic membrane exocytosis 3 | Rims3 |
A730060M23Rik, Nim3, Rim3, mKIAA0237 |
82.1kb | Anti-sense | Chr. 4 - 120854779 - 120896580 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |