Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAFAH1B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012095505.3 | XP_011950895.1 | 2 - 3 | 185 | 87 | |
XM_027979229.1 | XP_027835030.1 | 2 - 3 | 206 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |