Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162552.1 | NP_001156024.1 | 4 - 5 | 418 | 86 | |
GenBank mRNA | FJ969412.1 | - | 4 - 5 | 418 | 86 |