Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INHBA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009458.1 | NP_001009458.1 | 1 - 2 | 385 | 61 | |
XM_027968360.1 | XP_027824161.1 | 1 - 2 | 703 | 61 | ||
XM_027968361.1 | XP_027824162.1 | 3 - 4 | 921 | 61 | ||
XM_027968362.1 | XP_027824163.1 | 2 - 3 | 737 | 61 | ||
GenBank mRNA | L19218.1 | - | 1 - 2 | 385 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |