Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015092604.2 | XP_014948090.2 | 2 - 3 | 562 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |