Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UROD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012341.1 | NP_001012341.1 | 2 - 3 | 141 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |