Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004009638.4 | XP_004009687.1 | 2 - 3 | 965 | 114 | |
XM_012179547.3 | XP_012034937.1 | 2 - 3 | 2935 | 114 | ||
XM_012179552.3 | XP_012034942.1 | 2 - 3 | 2931 | 114 | ||
XM_027970825.1 | XP_027826626.1 | 2 - 3 | 1010 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |