Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009373.1 | NP_001009373.1 | 4 - 5 | 164 | 102 | |
GenBank mRNA | X16482.1 | - | 164 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |