Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GHSR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009760.1 | NP_001009760.1 | 1 - 2 | 862 | 67 | |
XM_027967424.1 | XP_027823225.1 | 2 - 3 | 1575 | 67 | ||
GenBank mRNA | AY093948.1 | - | 1 - 2 | 862 | 67 | |
AY093949.1 | - | 1 - 2 | 862 | 67 | ||
AY093950.1 | - | 1 - 2 | 862 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Recommended Assay Ovis aries |