Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRISP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185121.1 | NP_001172050.1 | 4 - 5 | 343 | 100 | |
GenBank mRNA | GU985271.1 | - | 4 - 5 | 343 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |