Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001308593.1 | NP_001295522.1 | 3 - 4 | 820 | 79 | |
XM_027978979.1 | XP_027834780.1 | 4 - 5 | 900 | 79 | ||
GenBank mRNA | JX534537.1 | - | 3 - 4 | 820 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |