Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFRP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027960330.1 | XP_027816131.1 | 1 - 2 | 530 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Ovis aries |