Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CATSPERB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027957272.1 | XP_027813073.1 | 25 - 26 | 3179 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |