Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMOD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012099691.3 | XP_011955081.1 | 2 - 3 | 1886 | 62 | |
XM_012099692.2 | XP_011955082.1 | 3 - 4 | 1925 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |